Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndufa4
Entrez Gene ID
406298
Gene Name
NADH:ubiquinone oxidoreductase subunit A4
Gene Aliases
zgc:73405
Chromosome Location
Chr.12: 104108 - 107620 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213025.1 NP_998190.1 2 - 3 139 79
GenBank mRNA BC059706.1 - 2 - 3 139 79

View Full Product Details