Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rbfox1l
Entrez Gene ID
407613
Gene Name
RNA binding fox-1 homolog 1, like
Gene Aliases
Fox-1, a2bp1l, fox1l, hm:zeh0082, zeh0082
Chromosome Location
Chr.16: 158466 - 176806 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214775.1 NP_999940.1 7 - 8 772 58
XM_005158173.4 XP_005158230.1 7 - 8 824 58
XM_021466671.1 XP_021322346.1 7 - 8 833 58
XM_021466672.1 XP_021322347.1 8 - 9 806 58
XM_021466673.1 XP_021322348.1 8 - 9 1055 58
GenBank mRNA AB074763.1 - 7 - 8 772 58
BC163738.1 - 7 - 8 851 58
BC163747.1 - 7 - 8 851 58

View Full Product Details