Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP6R3
Entrez Gene ID
100053490
Gene Name
protein phosphatase 6 regulatory subunit 3
Gene Aliases
SAPS3
Chromosome Location
Chr.12: 31467205 - 31607697 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113235.1 NP_001106706.1 22 - 23 2695 68
XM_023653812.1 XP_023509580.1 - 2709 68
XM_023653814.1 XP_023509582.1 - 2676 68
XM_023653816.1 XP_023509584.1 - 2640 68
XM_023653818.1 XP_023509586.1 - 2622 68
XM_023653821.1 XP_023509589.1 - 2589 68
GenBank mRNA AB371895.1 - 22 - 23 2695 68
AB371896.1 - 21 - 22 2608 68
AB371897.1 - 21 - 22 2590 68

View Full Product Details