Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA14
Entrez Gene ID
100462689
Gene Name
serine peptidase inhibitor clade A (alpha-1 antiproteinase, antitrypsin) member 14
Gene Aliases
-
Chromosome Location
Chr.24: 37509404 - 37519502 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242444.1 NP_001229373.1 1 - 2 653 88
XM_014735597.1 XP_014591083.1 - 746 88
GenBank mRNA EU810391.1 - 1 - 2 329 88

View Full Product Details