Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL18
Entrez Gene ID
421585
Gene Name
mitochondrial ribosomal protein L18
Gene Aliases
-
Chromosome Location
Chr.3: 45688638 - 45691180 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_419623.5 XP_419623.1 1 - 2 178 111
GenBank mRNA BX930290.1 - 1 - 2 150 111
BX930840.1 - 1 - 2 159 111
BX934122.1 - 1 - 2 184 111
BX935429.2 - 1 - 2 163 111
CR338829.2 - 1 - 2 159 111

View Full Product Details