Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3GALNT1
Entrez Gene ID
425011
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases
B3GALT3
Chromosome Location
Chr.9: 22252415 - 22276447 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015291773.2 XP_015147259.1 3 - 4 466 74
XM_015291775.2 XP_015147261.1 4 - 5 571 74
XM_025153491.1 XP_025009259.1 2 - 3 1218 74
XM_025153492.1 XP_025009260.1 2 - 3 1218 74
XM_025153493.1 XP_025009261.1 2 - 3 3811 74
XM_025153494.1 XP_025009262.1 5 - 6 692 74
XM_025153495.1 XP_025009263.1 4 - 5 550 74
XM_025153496.1 XP_025009264.1 2 - 3 3875 74
XM_025153497.1 XP_025009265.1 5 - 6 756 74
XM_025153498.1 XP_025009266.1 3 - 4 257 74
XM_025153499.1 XP_025009267.1 4 - 5 614 74
GenBank mRNA CR387037.1 - 4 - 5 554 74

View Full Product Details