Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NINJ2
Entrez Gene ID
418151
Gene Name
ninjurin 2
Gene Aliases
-
Chromosome Location
Chr.1: 60359916 - 60403894 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015286843.2 XP_015142329.2 3 - 4 523 80
GenBank mRNA CR406066.1 - 3 - 4 501 80

View Full Product Details