Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPO
Entrez Gene ID
776120
Gene Name
thyroid peroxidase
Gene Aliases
-
Chromosome Location
Chr.3: 92749569 - 92789803 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025148898.1 XP_025004666.1 2 - 3 251 99
XM_025148899.1 XP_025004667.1 2 - 3 232 99
GenBank mRNA BX933325.1 - 1 - 2 93 99

View Full Product Details