Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD50
Entrez Gene ID
422663
Gene Name
ankyrin repeat domain 50
Gene Aliases
-
Chromosome Location
Chr.4: 52874037 - 52916075 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015276388.2 XP_015131874.1 4 - 4 4337 85
GenBank mRNA BX929304.1 - 1 - 1 1353 85

View Full Product Details