Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKIG
Entrez Gene ID
419195
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor gamma
Gene Aliases
-
Chromosome Location
Chr.20: 5369169 - 5382842 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302151.1 NP_001289080.1 2 - 3 202 73
XM_025142192.1 XP_024997960.1 2 - 3 212 73
GenBank mRNA BG712998.1 - 2 - 3 202 73
BX929530.1 - 2 - 3 218 73

View Full Product Details