Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1GALT1
Entrez Gene ID
420575
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.2: 24688334 - 24702578 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012557.1 NP_001012575.1 2 - 3 355 117
XM_015281398.2 XP_015136884.1 2 - 3 410 117
XM_015281399.2 XP_015136885.1 2 - 3 257 117
GenBank mRNA AJ851683.1 - 2 - 3 355 117

View Full Product Details