Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSNK2A2
Entrez Gene ID
415626
Gene Name
casein kinase 2 alpha 2
Gene Aliases
-
Chromosome Location
Chr.11: 406954 - 429157 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012691.1 NP_001012709.1 9 - 10 903 76
XM_015292290.2 XP_015147776.1 9 - 10 862 76
XM_015292291.2 XP_015147777.1 8 - 9 951 76
GenBank mRNA M59457.1 - 9 - 10 903 76

View Full Product Details