Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KATNBL1
Entrez Gene ID
423287
Gene Name
katanin regulatory subunit B1 like 1
Gene Aliases
C5H15ORF29
Chromosome Location
Chr.5: 30227251 - 30242911 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031198.1 NP_001026369.1 7 - 8 779 112
XM_015287340.1 XP_015142826.1 7 - 8 1028 112
XM_015287341.2 XP_015142827.1 7 - 8 952 112
GenBank mRNA AJ720818.1 - 7 - 8 779 112

View Full Product Details