Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AvBD8
Entrez Gene ID
414875
Gene Name
avian beta-defensin 8
Gene Aliases
GAL8, Gal-12, Gal12
Chromosome Location
Chr.3: 107437978 - 107440736 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001781.1 NP_001001781.1 2 - 3 155 124
XM_015285097.2 XP_015140583.1 2 - 3 225 124
GenBank mRNA AY621310.1 - 2 - 3 155 124
AY701474.1 - 1 - 2 51 124
DQ677639.1 - 1 - 2 55 124
DQ858305.1 - 1 - 2 55 124
DQ858318.1 - 1 - 2 55 124
DQ858331.1 - 1 - 2 55 124
DQ858345.1 - 1 - 2 55 124

View Full Product Details