Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR3
Entrez Gene ID
374197
Gene Name
ARP3 actin related protein 3 homolog
Gene Aliases
-
Chromosome Location
Chr.7: 29611423 - 29637802 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204307.1 NP_989638.1 1 - 2 122 69
GenBank mRNA AF288600.1 - 1 - 2 122 69

View Full Product Details