Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PPIB
Entrez Gene ID
396447
Gene Name
peptidylprolyl isomerase B (cyclophilin B)
Gene Aliases
-
Chromosome Location
Chr.10: 1762529 - 1763819 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205461.1 NP_990792.1 1 - 2 110 70
GenBank mRNA M63553.1 - 1 - 2 110 70

View Full Product Details