Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P5CS
Entrez Gene ID
548058
Gene Name
delta-pyrroline-5-carboxylate synthetase
Gene Aliases
-
Chromosome Location
Chr.18: 2637348 - 2642823 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251224.1 NP_001238153.1 12 - 13 1438 76
GenBank mRNA AY492005.1 - 12 - 13 1438 76
FM999729.1 - 12 - 13 1438 76
FM999730.1 - 12 - 13 1438 76
FN564570.1 - 12 - 13 1438 76
FN564571.1 - 12 - 13 1438 76

View Full Product Details