Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGR2
Entrez Gene ID
732541
Gene Name
senescence-inducible chloroplast stay-green protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 53645859 - 53647879 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249761.1 NP_001236690.1 3 - 4 597 65
GenBank mRNA AY850142.1 - 3 - 4 597 65

View Full Product Details