Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098478.1 | NP_001091948.1 | 2 | 270 | |
NM_001098479.1 | NP_001091949.1 | 2 | 270 | ||
NM_018950.2 | NP_061823.2 | 2 | 270 | ||
XM_011514564.1 | XP_011512866.1 | 2 | 176 | ||
XM_017010810.1 | XP_016866299.1 | 2 | 176 | ||
XM_017010811.1 | XP_016866300.1 | 2 | 176 | ||
XM_017010812.1 | XP_016866301.1 | 2 | 176 | ||
XM_017010813.1 | XP_016866302.1 | 2 | 176 | ||
XM_017010814.1 | XP_016866303.1 | 2 | 157 | ||
XM_017010815.1 | XP_016866304.1 | 1 | 82 | ||
GenBank mRNA | AK096962.1 | - | 2 | 176 | |
AK125274.1 | - | 2 | 180 | ||
AK225185.1 | - | 2 | 174 | ||
AK298704.1 | - | 2 | 176 | ||
AK300090.1 | - | 2 | 209 | ||
AK301488.1 | - | 2 | 174 | ||
AK303670.1 | - | 1 | 306 | ||
AK309156.1 | - | 2 | 176 | ||
AY216682.1 | - | 2 | 134 | ||
AY221102.1 | - | 2 | 134 | ||
AY253269.1 | - | 2 | 146 | ||
AY253270.1 | - | 2 | 146 | ||
AY253271.1 | - | 2 | 146 | ||
BC009260.2 | - | 2 | 202 | ||
BC062991.1 | - | 2 | 150 | 5496696 | |
BM798587.1 | - | 2 | 271 | ||
CT002787.1 | - | 2 | 189 | ||
DQ367723.1 | - | 2 | 146 | ||
GQ891552.1 | - | 2 | 146 |