Gene Transcripts

Gene Symbol
MKLN1
Entrez Gene ID
4289
Gene Name
muskelin 1
Gene Aliases
TWA2
Chromosome Location
Chr. 7: 131110096 - 131496639 on Build GRCh38
UniGene ID
Hs.44693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145354.1 NP_001138826.1 9 964
NM_001321316.1 NP_001308245.1 8 895
NM_013255.4 NP_037387.2 8 892
XM_006715993.2 XP_006716056.1 8 892
XM_011516224.2 XP_011514526.1 8 892
GenBank mRNA AK002024.1 - 8 834
AK294384.1 - 9 965
AK310452.1 - 8 845
BC002834.2 - 3 270
BC067825.1 - 8 849 5268939

View Full Product Details