Gene Transcripts

Gene Symbol
MNDA
Entrez Gene ID
4332
Gene Name
myeloid cell nuclear differentiation antigen
Gene Aliases
PYHIN3
Chromosome Location
Chr. 1: 158831378 - 158849480 on Build GRCh38
UniGene ID
Hs.153837
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002432.1 NP_002423.1 5 1002
GenBank mRNA AK290392.1 - 5 1009
AK313210.1 - 5 1013
BC020715.1 - 5 1040
BC032319.1 - 5 1013 5223430
M81750.1 - 5 1002

View Full Product Details