Gene Transcripts

Gene Symbol
MTNR1A
Entrez Gene ID
4543
Gene Name
melatonin receptor 1A
Gene Aliases
MEL-1A-R, MT1
Chromosome Location
Chr. 4: 186532994 - 186555567 on Build GRCh38
UniGene ID
Hs.243467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005958.4 NP_005949.1 2 455
XM_011532002.2 XP_011530304.1 2 536
GenBank mRNA BC074946.2 - 2 271
BC074947.2 - 2 271 30915274
BC126297.1 - 2 455 8992013
BC126299.1 - 2 453 8992015
EU432127.1 - 2 253
U14108.1 - 2 285

View Full Product Details