Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001322019.1 | NP_001308948.1 | 3 | 424 | |
NM_001322020.1 | NP_001308949.1 | 3 | 424 | ||
NM_004544.3 | NP_004535.1 | 3 | 424 | ||
NR_136155.1 | 3 | 424 | |||
NR_136156.1 | 3 | 424 | |||
NR_136157.1 | 2 | 255 | |||
NR_136158.1 | 3 | 424 | |||
XM_011511228.2 | XP_011509530.1 | 3 | 411 | ||
GenBank mRNA | AB209440.1 | - | 3 | 345 | |
AF087661.1 | - | 3 | 344 | ||
AF087854.1 | - | 3 | 363 | ||
AF453834.1 | - | 3 | 323 | ||
AK290778.1 | - | 3 | 346 | ||
BC003417.1 | - | 3 | 358 | 3451514 | |
BC031332.1 | - | 3 | 329 | 4872196 | |
BI551914.1 | - | 408 |