Gene Transcripts

Gene Symbol
NTSR1
Entrez Gene ID
4923
Gene Name
neurotensin receptor 1 (high affinity)
Gene Aliases
NTR
Chromosome Location
Chr. 20: 62708770 - 62762771 on Build GRCh38
UniGene ID
Hs.590869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002531.2 NP_002522.2 3, 4 1379
GenBank mRNA AY429106.1 - 3, 4 1008
X70070.1 - 3, 4 1380

View Full Product Details