Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001312673.1 | NP_001299602.1 | 4 | 441 | |
NM_005017.3 | NP_005008.2 | 5 | 505 | ||
GenBank mRNA | AK304534.1 | - | 4 | 298 | |
AK309073.1 | - | 4 | 441 | ||
AK309300.1 | - | 4 | 446 | ||
AK313450.1 | - | 4 | 363 | ||
BC046355.1 | - | 5 | 466 | 5744747 | |
L28957.1 | - | 4 | 338 |