Gene Transcripts
- Gene Symbol
- SEPT4
- Entrez Gene ID
- 5414
- Gene Name
- septin 4
- Gene Aliases
- ARTS, BRADEION, CE5B3, H5, MART, PNUTL2, SEP4, hCDCREL-2, hucep-7
- Chromosome Location
- Chr. 17: 58520250 - 58540818 on Build GRCh38
- UniGene ID
- Hs.287518
- Species
- Homo sapiens
View Full Product Details