Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002865.2 | NP_002856.1 | 2, 3 | 407 | |
GenBank mRNA | AF498930.1 | - | 2, 3 | 109 | |
AK312344.1 | - | 2, 3 | 330 | ||
BC008929.2 | - | 2, 3 | 297 | 2966694 | |
BT019695.1 | - | 2, 3 | 109 | ||
DA648682.1 | - | 2, 3 | 409 | ||
M28213.1 | - | 2, 3 | 149 | ||
X12953.1 | - | 2, 3 | 317 | ||
X13899.1 | - | 2, 3 | 109 |