Gene Transcripts

Gene Symbol
TACR2
Entrez Gene ID
6865
Gene Name
tachykinin receptor 2
Gene Aliases
NK2R, NKNAR, SKR, TAC2R
Chromosome Location
Chr. 10: 69404202 - 69416918 on Build GRCh38
UniGene ID
Hs.88372
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001057.2 NP_001048.2 4 1447
GenBank mRNA AK096906.1 - 2 472
AK291996.1 - 4 1447
AK300649.1 - 4 1245
AY322545.1 - 4 852
BC096842.1 - 4 858 6971906
FJ200479.1 - 5 744
M57414.1 - 4 852

View Full Product Details