Gene Transcripts

Gene Symbol
TPST2
Entrez Gene ID
8459
Gene Name
tyrosylprotein sulfotransferase 2
Gene Aliases
TANGO13B
Chromosome Location
Chr. 22: 26524082 - 26590125 on Build GRCh38
UniGene ID
Hs.632768
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008566.1 NP_001008566.1 3 810
NM_003595.3 NP_003586.3 3 702
XM_006724338.3 XP_006724401.1 4 821
GenBank mRNA AF049891.1 - 3 666
AF061254.1 - 1 474
AJ006198.1 - 1 556
AK074538.1 - 3 702
AK075139.1 - 3 774
AK304545.1 - 5 786
AL136623.2 - 3 686
BC001057.2 - 3 681 3509627
BC017509.1 - 3 666 4857366
CR456597.1 - 1 564
CR533528.1 - 1 469

View Full Product Details