Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145713.1 | NP_001139185.1 | 4 | 568 | |
NM_001145714.1 | NP_001139186.1 | 4 | 568 | ||
NM_005500.2 | NP_005491.1 | 4 | 568 | ||
NR_027280.1 | 4 | 617 | |||
XM_006722963.2 | XP_006723026.1 | 4 | 568 | ||
XM_006722965.3 | XP_006723028.1 | 4 | 568 | ||
XM_017026135.1 | XP_016881624.1 | 4 | 568 | ||
XM_017026136.1 | XP_016881625.1 | 4 | 439 | ||
GenBank mRNA | AF046025.2 | - | 4 | 505 | |
AF090385.2 | - | 4 | 513 | ||
AF110956.1 | - | 4 | 460 | ||
AK021978.1 | - | 4 | 562 | ||
AK027595.1 | - | 4 | 532 | ||
AK055481.1 | - | 4 | 533 | ||
AK090645.1 | - | 4 | 586 | ||
AK315624.1 | - | 4 | 510 | ||
BC000344.2 | - | 4 | 521 | 2823028 | |
BC003611.2 | - | 4 | 489 | 2988252 | |
BC018271.1 | - | 4 | 528 | 3452340 | |
BT007290.1 | - | 4 | 460 |