Gene Transcripts
- Gene Symbol
- ARPC2
- Entrez Gene ID
- 10109
- Gene Name
- actin related protein 2/3 complex subunit 2
- Gene Aliases
- ARC34, PNAS-139, PRO2446, p34-Arc
- Chromosome Location
- Chr. 2: 218217094 - 218254348 on Build GRCh38
- UniGene ID
- Hs.529303
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005731.3 |
NP_005722.1
|
4, 5 |
336 |
|
NM_152862.2 |
NP_690601.1
|
5, 6 |
431 |
|
XM_017003113.1 |
XP_016858602.1
|
4, 5 |
305 |
|
GenBank mRNA |
AF006085.1 |
- |
4, 5 |
346 |
|
AK092520.1 |
- |
1, 2 |
1393 |
|
AK290268.1 |
- |
5, 6 |
330 |
|
BC000590.2 |
- |
5, 6 |
294 |
3346685 |
BG282181.1 |
- |
5, 6 |
430 |
|
BG765898.1 |
- |
5, 6 |
374 |
|
BT006898.1 |
- |
4, 5 |
262 |
|
U50523.1 |
- |
5, 6 |
346 |
|
View Full Product Details