Gene Transcripts
- Gene Symbol
- TRIM2
- Entrez Gene ID
- 23321
- Gene Name
- tripartite motif containing 2
- Gene Aliases
- CMT2R, RNF86
- Chromosome Location
- Chr. 4: 153152281 - 153339322 on Build GRCh38
- UniGene ID
- Hs.435711
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130067.1 |
NP_001123539.1
|
6, 7 |
1635 |
|
NM_015271.4 |
NP_056086.2
|
6, 7 |
1632 |
|
XM_006714157.1 |
XP_006714220.1
|
6, 7 |
1632 |
|
XM_006714158.1 |
XP_006714221.1
|
6, 7 |
1597 |
|
XM_006714159.3 |
XP_006714222.1
|
6, 7 |
2443 |
|
XM_006714160.1 |
XP_006714223.1
|
7, 8 |
2397 |
|
XM_006714161.1 |
XP_006714224.1
|
7, 8 |
2394 |
|
XM_006714162.1 |
XP_006714225.1
|
7, 8 |
2114 |
|
XM_006714165.3 |
XP_006714228.1
|
4, 5 |
1825 |
|
XM_011531794.1 |
XP_011530096.1
|
6, 7 |
1711 |
|
XM_011531795.1 |
XP_011530097.1
|
7, 8 |
2117 |
|
XM_011531796.2 |
XP_011530098.1
|
6, 7 |
1647 |
|
XM_011531798.2 |
XP_011530100.1
|
6, 7 |
1752 |
|
XM_017007943.1 |
XP_016863432.1
|
6, 7 |
1597 |
|
XM_017007944.1 |
XP_016863433.1
|
7, 8 |
2397 |
|
XM_017007945.1 |
XP_016863434.1
|
7, 8 |
2394 |
|
XM_017007946.1 |
XP_016863435.1
|
6, 7 |
2440 |
|
XM_017007947.1 |
XP_016863436.1
|
6, 7 |
1711 |
|
XM_017007948.1 |
XP_016863437.1
|
6, 7 |
1665 |
|
XM_017007949.1 |
XP_016863438.1
|
7, 8 |
2114 |
|
XM_017007950.1 |
XP_016863439.1
|
7, 8 |
2117 |
|
XM_017007951.1 |
XP_016863440.1
|
6, 7 |
1811 |
|
XM_017007952.1 |
XP_016863441.1
|
6, 7 |
1819 |
|
XM_017007953.1 |
XP_016863442.1
|
4, 5 |
1163 |
|
XM_017007954.1 |
XP_016863443.1
|
7, 8 |
1689 |
|
XM_017007955.1 |
XP_016863444.1
|
7, 8 |
1682 |
|
GenBank mRNA |
AB011089.1 |
- |
6, 7 |
1580 |
|
AF220018.1 |
- |
6, 7 |
1580 |
|
BC005016.1 |
- |
1, 2 |
152 |
|
BC011052.1 |
- |
6, 7 |
1635 |
4156234 |
View Full Product Details