Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012476.2 | NP_036608.1 | 2 | 292 | |
XM_006711982.3 | XP_006712045.1 | 2 | 331 | ||
XM_011532750.2 | XP_011531052.1 | 2 | 331 | ||
XM_011532751.2 | XP_011531053.1 | 2 | 331 | ||
GenBank mRNA | AB593145.1 | - | 2 | 301 | |
BC006336.2 | - | 2 | 267 | 4304066 | |
BT007035.1 | - | 2 | 260 | ||
Y17791.1 | - | 2 | 292 |