Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271814.1 | NP_001258743.1 | 2 | 143 | |
NM_012400.3 | NP_036532.1 | 2 | 143 | ||
GenBank mRNA | AF112982.1 | - | 2 | 103 | |
AF188625.1 | - | 2 | 111 | ||
AK290406.1 | - | 2 | 104 | ||
AK301396.1 | - | 2 | 143 | ||
AK310156.1 | - | 2 | 113 | ||
AK311903.1 | - | 2 | 113 | ||
BC025706.1 | - | 2 | 92 | 5223912 | |
DC295429.1 | - | 2 | 143 |