Gene Transcripts

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
Chromosome Location
Chr. 11: 86302211 - 86345943 on Build GRCh38
UniGene ID
Hs.283322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322404.1 NP_001309333.1 3 595
NM_001322407.1 NP_001309336.1 4 769
NM_001322409.1 NP_001309338.1 4 699
NM_016401.3 NP_057485.2 3 611
NR_024597.1 2 373
NR_024598.1 2 373
NR_136324.1 4 719
XM_017017914.1 XP_016873403.1 3 601
XM_017017915.1 XP_016873404.1 3 388
GenBank mRNA AA148558.1 - 2 338
AF151082.1 - 2 254
AF161487.1 - 3 592
AF161527.1 - 3 595
AK001447.1 - 3 582
AK223198.1 - 3 582
AK298763.1 - 4 735
BC001677.2 - 3 484 2820772
BC006476.2 - 3 484 2820772
BC015991.2 - 3 576 2820691
BC018080.1 - 3 545 4798958
BC021621.1 - 3 576 2820691
BE858273.1 - 4 347
BP423222.1 - 2 290
BX332668.2 - 3 448

View Full Product Details