Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271762.1 | NP_001258691.1 | 16 | 1851 | |
NM_001271763.1 | NP_001258692.1 | 15 | 1826 | ||
NM_017803.4 | NP_060273.1 | 17 | 1931 | ||
GenBank mRNA | AB101210.1 | - | 17 | 1595 | |
AK000406.1 | - | 16 | 1498 | ||
AK290578.1 | - | 17 | 1633 | ||
AK296663.1 | - | 15 | 1478 | ||
BC006527.2 | - | 17 | 1574 | 2988128 |