Gene Transcripts

Gene Symbol
TTC12
Entrez Gene ID
54970
Gene Name
tetratricopeptide repeat domain 12
Gene Aliases
TPARM
Chromosome Location
Chr. 11: 113314529 - 113385952 on Build GRCh38
UniGene ID
Hs.288772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318533.1 NP_001305462.1 20 1923
NM_017868.3 NP_060338.3 20 1905
XM_005271604.3 XP_005271661.1 20 2121
XM_011542890.2 XP_011541192.1 21 2119
XM_017017943.1 XP_016873432.1 20 1905
XM_017017944.1 XP_016873433.1 21 2475
XM_017017945.1 XP_016873434.1 21 1958
XM_017017946.1 XP_016873435.1 20 2128
XM_017017947.1 XP_016873436.1 20 1905
XM_017017948.1 XP_016873437.1 20 1905
XM_017017949.1 XP_016873438.1 20 1905
XM_017017950.1 XP_016873439.1 20 1905
GenBank mRNA AK000542.1 - 19 1797
AK023921.1 - 20 1851
AK091887.1 - 22 2517
AK223117.1 - 19 1797
AK292331.1 - 20 1843
BC011546.1 - 19 1847
BC032355.1 - 19 1827 5219117

View Full Product Details