Gene Transcripts
- Gene Symbol
- ZDHHC7
- Entrez Gene ID
- 55625
- Gene Name
- zinc finger DHHC-type containing 7
- Gene Aliases
- DHHC7, SERZ-B, SERZ1, ZNF370
- Chromosome Location
- Chr. 16: 84974460 - 85011732 on Build GRCh38
- UniGene ID
- Hs.461610
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145548.1 |
NP_001139020.1
|
5, 6 |
900 |
|
NM_017740.2 |
NP_060210.2
|
4, 5 |
789 |
|
XM_017023434.1 |
XP_016878923.1
|
2, 3 |
325 |
|
XM_017023435.1 |
XP_016878924.1
|
2, 3 |
185 |
|
XM_017023436.1 |
XP_016878925.1
|
2, 3 |
300 |
|
GenBank mRNA |
AK000286.1 |
- |
4, 5 |
789 |
|
AK001654.1 |
- |
2, 3 |
175 |
|
AK225768.1 |
- |
4, 5 |
741 |
|
BC017702.1 |
- |
2, 3 |
150 |
4394921 |
BC018772.1 |
- |
5, 6 |
887 |
4869114 |
View Full Product Details