Gene Transcripts
- Gene Symbol
- PLEKHA4
- Entrez Gene ID
- 57664
- Gene Name
- pleckstrin homology domain containing A4
- Gene Aliases
- PEPP1
- Chromosome Location
- Chr. 19: 48837092 - 48868627 on Build GRCh38
- UniGene ID
- Hs.9469
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161354.1 |
NP_001154826.1
|
9, 10 |
1571 |
|
NM_020904.2 |
NP_065955.2
|
10, 11 |
1646 |
|
XM_005259107.2 |
XP_005259164.1
|
9, 10 |
1566 |
|
XM_006723301.2 |
XP_006723364.1
|
9, 10 |
1566 |
|
XM_011527157.1 |
XP_011525459.1
|
10, 11 |
1653 |
|
XM_011527158.2 |
XP_011525460.1
|
9, 10 |
1492 |
|
XM_011527159.1 |
XP_011525461.1
|
10, 11 |
1666 |
|
XM_011527160.1 |
XP_011525462.1
|
10, 11 |
1653 |
|
XM_011527162.2 |
XP_011525464.1
|
3, 4 |
262 |
|
XM_017027037.1 |
XP_016882526.1
|
7, 8 |
930 |
|
GenBank mRNA |
AB208908.1 |
- |
10, 11 |
1608 |
|
AY007233.1 |
- |
10, 11 |
1620 |
|
BC024157.2 |
- |
10, 11 |
1626 |
3953478 |
BC033832.1 |
- |
2, 3 |
96 |
|
BC064601.1 |
- |
9, 10 |
1398 |
5785991 |
View Full Product Details