Gene Transcripts

Gene Symbol
LSM2
Entrez Gene ID
57819
Gene Name
LSM2 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
C6orf28, G7B, YBL026W, snRNP
Chromosome Location
Chr. 6: 31797392 - 31806984 on Build GRCh38
UniGene ID
Hs.103106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021177.4 NP_067000.1 5 459
GenBank mRNA AF136977.1 - 5 444
AF182288.1 - 5 278
AF196468.1 - 5 424
AI570628.1 - 1 419
AJ245416.1 - 5 450
BC009192.2 - 5 441 3842537
BP203102.1 - 5 459
BU189898.1 - 5 450
CR542146.1 - 4 232
CR542157.1 - 4 232

View Full Product Details