Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166621.1 | NP_001160093.1 | 5 | 456 | |
NM_021210.4 | NP_067033.1 | 4 | 516 | ||
NR_030684.1 | 3 | 445 | |||
NR_030697.1 | 4 | 385 | |||
GenBank mRNA | AK314038.1 | - | 4 | 522 | |
BC025413.1 | - | 4 | 375 | ||
BC032717.1 | - | 4 | 518 | 5518603 | |
BI457771.1 | - | 4 | 518 | ||
BI600633.1 | - | 5 | 461 | ||
BM762006.1 | - | 5 | 430 | ||
BU687854.1 | - | 1 | 319 | ||
BU940685.1 | - | 4 | 351 |