Gene Transcripts
- Gene Symbol
- UPF3A
- Entrez Gene ID
- 65110
- Gene Name
- UPF3 regulator of nonsense transcripts homolog A (yeast)
- Gene Aliases
- HUPF3A, RENT3A, UPF3
- Chromosome Location
- Chr. 13: 114281584 - 114305817 on Build GRCh38
- UniGene ID
- Hs.533855
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023011.3 |
NP_075387.1
|
5, 6 |
690 |
|
NM_080687.2 |
NP_542418.1
|
4, 5 |
591 |
|
XM_005266202.4 |
XP_005266259.1
|
2, 3 |
316 |
|
XM_006719991.3 |
XP_006720054.1
|
4, 5 |
565 |
|
XM_011534845.2 |
XP_011533147.1
|
5, 6 |
1218 |
|
XM_011534846.1 |
XP_011533148.1
|
5, 6 |
637 |
|
XM_011534847.2 |
XP_011533149.1
|
4, 5 |
1030 |
|
XM_011534848.2 |
XP_011533150.1
|
5, 6 |
1175 |
|
XM_017020709.1 |
XP_016876198.1
|
4, 5 |
1402 |
|
XM_017020710.1 |
XP_016876199.1
|
5, 6 |
787 |
|
XM_017020711.1 |
XP_016876200.1
|
4, 5 |
688 |
|
GenBank mRNA |
AF318575.1 |
- |
5, 6 |
583 |
|
AK097027.1 |
- |
1, 2 |
652 |
|
AK302003.1 |
- |
5, 6 |
626 |
|
AY013250.1 |
- |
5, 6 |
690 |
|
BC023569.1 |
- |
4, 5 |
555 |
4127465 |
View Full Product Details