Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025205.4 | NP_079481.2 | 1 | 156 | |
GenBank mRNA | AF317678.1 | - | 1 | 128 | |
AF317679.1 | - | 1 | 133 | ||
AF317680.1 | - | 1 | 140 | ||
AF318059.1 | - | 1 | 153 | ||
AF358829.1 | - | 1 | 141 | ||
AK313928.1 | - | 1 | 138 | ||
BC011936.2 | - | 1 | 133 | 3936713 | |
HY174042.1 | - | 1 | 156 |