Gene Transcripts

Gene Symbol
PITPNM3
Entrez Gene ID
83394
Gene Name
PITPNM family member 3
Gene Aliases
ACKR6, CORD5, NIR1, RDGBA3
Chromosome Location
Chr. 17: 6451263 - 6556557 on Build GRCh38
UniGene ID
Hs.183983
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165966.1 NP_001159438.1 5 608
NM_031220.3 NP_112497.2 6 716
XM_011524015.2 XP_011522317.1 6 656
XM_011524016.2 XP_011522318.1 6 657
XM_011524017.2 XP_011522319.1 6 658
GenBank mRNA AF334586.1 - 6 713
AL389994.1 - 5 439
BC128583.1 - 6 653 40128716
BC128584.1 - 6 653 40128724

View Full Product Details