Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_017020002.1 | XP_016875491.1 | 2 | 657 | |
GenBank mRNA | AF212239.1 | - | 2 | 269 | |
AK096947.1 | - | 1 | 568 | ||
AK223378.1 | - | 2 | 284 | ||
AK311977.1 | - | 2 | 282 | ||
AY007436.1 | - | 2 | 265 | ||
BC029355.1 | - | 2 | 286 | 4613026 |