Gene Transcripts
- Gene Symbol
- C16orf89
- Entrez Gene ID
- 146556
- Gene Name
- chromosome 16 open reading frame 89
- Gene Aliases
- Chromosome Location
- Chr. 16: 5042772 - 5066145 on Build GRCh38
- UniGene ID
- Hs.11782
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001098514.2 |
NP_001091984.2
|
1, 2 |
441 |
|
NM_152459.4 |
NP_689672.4
|
1, 2 |
441 |
|
XM_005255143.3 |
XP_005255200.2
|
1, 2 |
267 |
|
XM_017022972.1 |
XP_016878461.1
|
1, 2 |
267 |
|
XM_017022973.1 |
XP_016878462.1
|
1, 2 |
267 |
|
XM_017022974.1 |
XP_016878463.1
|
1, 2 |
267 |
|
GenBank mRNA |
AK075242.1 |
- |
1, 2 |
267 |
|
AK300708.1 |
- |
1, 2 |
215 |
|
AY358483.1 |
- |
1, 2 |
406 |
|
BC033681.1 |
- |
1, 2 |
336 |
5190553 |
View Full Product Details