Gene Transcripts

Gene Symbol
MCOLN2
Entrez Gene ID
255231
Gene Name
mucolipin 2
Gene Aliases
TRP-ML2, TRPML2
Chromosome Location
Chr. 1: 84925583 - 84997122 on Build GRCh38
UniGene ID
Hs.591446
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153259.3 NP_694991.2 11 1547
XM_005270719.3 XP_005270776.1 11 1324
XM_006710552.2 XP_006710615.1 12 1637
XM_011541187.2 XP_011539489.1 11 1381
XM_011541188.2 XP_011539490.1 11 1664
GenBank mRNA AK094010.1 - 9 1222
AK129786.1 - 5 550
AK309858.1 - 11 1538
AY083533.1 - 10 1213
BC104891.1 - 11 1365 8143894
BC104893.1 - 11 1365 8143896

View Full Product Details