Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152772.2 | NP_689985.1 | 8 | 1240 | |
XM_005268767.4 | XP_005268824.1 | 9 | 1390 | ||
XM_005268768.4 | XP_005268825.1 | 8 | 1396 | ||
XM_011538129.2 | XP_011536431.1 | 9 | 2158 | ||
XM_011538130.2 | XP_011536432.1 | 9 | 1535 | ||
XM_011538131.2 | XP_011536433.1 | 9 | 1393 | ||
XM_017019127.1 | XP_016874616.1 | 8 | 2069 | ||
XM_017019128.1 | XP_016874617.1 | 10 | 2305 | ||
XM_017019129.1 | XP_016874618.1 | 10 | 1793 | ||
XM_017019130.1 | XP_016874619.1 | 9 | 1341 | ||
XM_017019131.1 | XP_016874620.1 | 8 | 1113 | ||
XM_017019132.1 | XP_016874621.1 | 10 | 1532 | ||
XM_017019133.1 | XP_016874622.1 | 7 | 1060 | ||
GenBank mRNA | AK314055.1 | - | 8 | 1180 | |
BC033617.1 | - | 8 | 1228 | 5248900 |