Gene Transcripts
- Gene Symbol
- LOC107983965
- Entrez Gene ID
- 107983965
- Gene Name
- orofacial cleft 1 candidate gene 1 protein-like
- Gene Aliases
- Chromosome Location
- Chr. 6: 9850416 - 10211608 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- OFCC1
- Entrez Gene ID
- 266553
- Gene Name
- orofacial cleft 1 candidate 1
- Gene Aliases
- MRDS1
- Chromosome Location
- Chr. 6: 9775808 - 9842630 on Build GRCh38
- UniGene ID
- Hs.532138
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF520800.1 |
- |
3, 4 |
162 |
|
AF520803.1 |
- |
1, 2 |
34 |
|
AF520804.1 |
- |
1, 2 |
20 |
|
AF520805.1 |
- |
1, 2 |
34 |
|
AF520806.1 |
- |
1, 2 |
42 |
|
AF520807.1 |
- |
2, 3 |
92 |
|
AF520808.1 |
- |
2, 3 |
122 |
|
AF548113.1 |
- |
3, 4 |
308 |
|
AY309094.1 |
- |
4, 5 |
243 |
|
BC113541.1 |
- |
2, 3 |
101 |
8322593 |
BC113543.1 |
- |
2, 3 |
101 |
8322595 |
BC143837.1 |
- |
2, 3 |
101 |
9052352 |
View Full Product Details