Gene Transcripts

Gene Symbol
OR1J1
Entrez Gene ID
347168
Gene Name
olfactory receptor family 1 subfamily J member 1
Gene Aliases
OR9-18, hg32
Chromosome Location
Chr. 9: 122476958 - 122477926 on Build GRCh38
UniGene ID
Hs.632692
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004451.1 NP_001004451.1 1 382
GenBank mRNA BC136933.1 - 1 412
BC136938.1 - 1 412 9020930

View Full Product Details